Learn More
New Jersey Fluid Systems LLC Swagelok HC-200-7-4 Female Connector, Alloy C-276, Leak-Tight Connection, 1/8 in Tube OD to 1/4 in Female NPT
Catalog No. NC3714508
Encompass
Introducing the Swagelok HC-200-7-4 Female Connector, designed for unmatched reliability in demanding environments. Crafted from Alloy C-276, this connector provides superior corrosion resistance and ensures a secure, leak-tight connection between 1/8 in tube OD and 1/4 in female NPT. Its innovative two-ferrule design simplifies installation while maintaining integrity under pressure, making it ideal for critical fluid systems. Enjoy confidence in performance and longevity with this robust solution.
Catalog No. NC3714508 Supplier New Jersey Fluid Systems LLC Supplier No. HC20074
Introducing the Swagelok HC-200-7-4 Female Connector, designed for unmatched reliability in demanding environments. Crafted from Alloy C-276, this connector provides superior corrosion resistance and ensures a secure, leak-tight connection between 1/8 in tube OD and 1/4 in female NPT. Its innovative two-ferrule design simplifies installation while maintaining integrity under pressure, making it ideal for critical fluid systems. Enjoy confidence in performance and longevity with this robust solution.