Learn More
TARGETMOL CHEMICALS INC C29 10MG
SDP
Catalog No. 502580237
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. C29 is a novel inhibitor of TLR2/1 and TLR2/6 signaling induced by synthetic and bacterial TLR2 agonists in human HEK-TLR2 and THP-1 cells but only TLR2/1 signaling in murine macrophages. purity: 98%
Catalog No. 50-258-0237 Supplier TARGETMOL CHEMICALS INC Supplier No. T350210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. C29 is a novel inhibitor of TLR2/1 and TLR2/6 signaling induced by synthetic and bacterial TLR2 agonists in human HEK-TLR2 and THP-1 cells but only TLR2/1 signaling in murine macrophages. purity: 98%