Learn More
Eurofins Discoverx PathHunter C2C12 CXCR5 β-Arrestin Cell Line, 2 vials. (93-0204C7)
Catalog No. 501999940
Encompass
The PathHunter CXCR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. [ligand]) based activation of CXCR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR5 and EA-tagged β-Arrestin-2. Activation of CXCR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9940 Supplier Eurofins Discoverx Supplier No. 930204C7
Add to Cart
Add to Cart
Request a Quote
The PathHunter CXCR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. [ligand]) based activation of CXCR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR5 and EA-tagged β-Arrestin-2. Activation of CXCR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.