Learn More
ATCC C2C12; Muscle Myoblast; Mouse (Mus musculus)
Catalog No. 502382000
Encompass
C2C12 is a myoblast cell line that is a subclone (produced by H. Blau, et al) of the mouse myoblast cell line established by D. Yaffe and O. Saxel. The C2C12 cell line differentiates rapidly, forming contractile myotubes and producing characteristic muscle proteins. Treatment with bone morphogenic protein 2 (BMP-2) cause a shift in the differentiation pathway from myoblastic to osteoblastic.
Catalog No. 50-238-2000 Supplier ATCC Supplier No. CRL1772
 Restricted Item
Add to Cart
Add to Cart
C2C12 is a myoblast cell line that is a subclone (produced by H. Blau, et al) of the mouse myoblast cell line established by D. Yaffe and O. Saxel. The C2C12 cell line differentiates rapidly, forming contractile myotubes and producing characteristic muscle proteins. Treatment with bone morphogenic protein 2 (BMP-2) cause a shift in the differentiation pathway from myoblastic to osteoblastic.