Learn More
ABclonal Technology C2CD5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143571000UL
Encompass_Preferred
Enables calcium ion binding activity and calcium-dependent phospholipid binding activity. Involved in cellular response to insulin stimulus, intracellular protein transmembrane transport, and positive regulation of transport. Located in several cellular components, including centriolar satellite, cytoplasmic vesicle membrane, and ruffle membrane.
Catalog No. 50-001-07030