Learn More
Aviva Systems Biology Corp C2orf18 Antibody - middle region (ARP47368_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47368_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C2orf18. Alternate names: FLJ20555, ANT2BP, C2orf18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59525