Learn More
Aviva Systems Biology Corp C2orf28 Antibody - C-terminal region (ARP36564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf28. It was validated on Western Blot. . Alternate names: APR--3, APR-3, APR3, HSPC013, PRO240, p18, C2orf28ApplicationWB. Species reactivity: Human, Pig, Rabbit, Rat
Catalog No. 50-284-502