Learn More
Aviva Systems Biology Corp C2orf28 Antibody - C-terminal region (ARP30550_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C2orf28. Alternate names: APR--3, APR-3, APR3, HSPC013, PRO240, p18, C2orf28ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54162