Learn More
Aviva Systems Biology Corp C2orf3 Antibody - N-terminal region (ARP58090_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58090_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C2orf3. Alternate names: DNABF, GCF, TCF9, C2orf3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62233