Learn More
Aviva Systems Biology Corp C2orf33 Antibody - C-terminal region (ARP49483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C2orf33. Alternate names: DKFZp666J168, GL004, MGC110913, C2orf33ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60093