Learn More
Aviva Systems Biology Corp LDAH Antibody - C-terminal region (ARP62773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf43. It was validated on Western Blot. . Alternate names: FLJ21820ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-766