Learn More
Aviva Systems Biology Corp CATIP Antibody - C-terminal region (ARP55902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf62. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human C2orf62. Alternate names: MGC50811ApplicationWB. Species reactivity: Cow, Human, Rabbit, Rat
Catalog No. 50-286-094