Description
- Sevoflurane, Key Fill
- Standard capacity, 125 mL
- Quality materials
- Brass, stainless steel
- Advanced wicking system
- Calibrated using laser refractometry assures accuracy
- Safe, leak proof performance
- One piece fill system
Specifications
Specifications
| For Use With (Application) | Animal Inhalant Anesthesia |
| Animal | All Animals |
| Type | Anesthetic Vaporizer |