Learn More
Abcam Human Complement C3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088231X96TESTS
Encompass_Preferred
Human Complement C3 ELISA Kit is a Sandwich ELISA used to quantify Human Complement C3 with a sensitivity of 84 pg/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Wide dynamic range – 0.625 - 40 ng/ml - Cited in over 15 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0747282