Learn More
Cayman Chemical C30 Ceramide (d18:1/30:0), Purity: ≥98%, CAS: 871540-97-5, 1 mg
SDP
Catalog No. 502936124
Encompass_Preferred
A naturally occurring ceramide; has been found in the stratum corneum of human and nude mouse skin; decreased in the skin of patients with lesional and non-lesional atopic dermatitis
Catalog No. 50-293-6124 Supplier Cayman Chemical Supplier No. 244001 MG
A naturally occurring ceramide; has been found in the stratum corneum of human and nude mouse skin; decreased in the skin of patients with lesional and non-lesional atopic dermatitis