Learn More
Cayman Chemical C30 (ω-hydroxy) Ceramide (d18:1/30:0), Purity: ≥98%, CAS: 457100-08-2, 5 mg
SDP
Catalog No. 502995525
Encompass_Preferred
A bioactive sphingolipid vital to skin barrier functions; deficiency causes defective skin-water permeability barrier function and neonatality in ELOVL4 deficient mice
Catalog No. 50-299-5525 Supplier Cayman Chemical Supplier No. 226885 MG
A bioactive sphingolipid vital to skin barrier functions; deficiency causes defective skin-water permeability barrier function and neonatality in ELOVL4 deficient mice