Learn More
Cayman Chemical C32 CeramIded18 132 0 5mg
SDP
Catalog No. 502935931
Encompass_Preferred
A naturally occurring ceramide; has been found in the stratum corneum of human and nude mouse skin
Catalog No. 50-293-5931 Supplier Cayman Chemical Supplier No. 244245 MG
A naturally occurring ceramide; has been found in the stratum corneum of human and nude mouse skin