Learn More
ATCC C-33 A; Cervical Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB31
Encompass
C-33 A [c-33a] are epithelial cells isolated from the cervix of a 66-year-old, White, uterine cancer patient. Expression of p53+ is elevated, and there is a point mutation at codon 273 resulting in a Arg -> Cys substitution.
Catalog No. 50-238-4396