Learn More
Alkali Scientific Alkali Scientific™ C3303-6HP Sprint™ 6H Plus Clinical Lab Centrifuge With 6 x 15mL Swing-Out Rotor
SDP
Catalog No. 503029677
Encompass_Preferred
Alkali Scientific™ C3303-6HP Sprint 6H Plus is our premium centrifuge offering providinghorizontal centrifuge: – Increased speed up to 4,000rpm / 2504xg– Back-lit, liquid crystal display– Adjustable speed in increments of 100rpm– Digital speed control in rpm or rcf– Adjustable acceleration and deceleration levels– A safety imbalance detection system– Momentary operation– High capacity rotor for tubes up to 15ml (conical or round bottom) In addition, the Sprint 6H PLUS offers an optional fixed-angle rotor forcentrifugation of up to 12 tubes. Ideal for Coag, PPP and urinalysis studies
Catalog No. 50-302-9677 Supplier Alkali Scientific Supplier No. RS7533
Alkali Scientific™ C3303-6HP Sprint 6H Plus is our premium centrifuge offering providinghorizontal centrifuge: – Increased speed up to 4,000rpm / 2504xg– Back-lit, liquid crystal display– Adjustable speed in increments of 100rpm– Digital speed control in rpm or rcf– Adjustable acceleration and deceleration levels– A safety imbalance detection system– Momentary operation– High capacity rotor for tubes up to 15ml (conical or round bottom) In addition, the Sprint 6H PLUS offers an optional fixed-angle rotor forcentrifugation of up to 12 tubes. Ideal for Coag, PPP and urinalysis studies