Learn More
MSC Starrett 56262 5-3/4" Long, 1/50, 1/10" and 0.5, 1mm Graduation, Flexible Steel Rule Decimal/Metric Graduation Style, 1/2" Wide, Silver, Satin Chrome Finish
Catalog No. 501304778
Encompass
Starrett 56262 5-3/4" Long, 1/50, 1/10" and 0.5, 1mm Graduation, Flexible Steel Rule Decimal/Metric Graduation Style, 1/2" Wide, Silver, Satin Chrome Finish
Catalog No. 50-130-4778 Supplier MSC Supplier No. 86413077
Starrett 56262 5-3/4" Long, 1/50, 1/10" and 0.5, 1mm Graduation, Flexible Steel Rule Decimal/Metric Graduation Style, 1/2" Wide, Silver, Satin Chrome Finish