Learn More
Strem, An Ascensus Company CAS# 33309-88-5. Bis(tricyclohexylphosphine)palladium(0), 98%. MFCD01073796. 5G.
SDP
Catalog No. 501959577
Encompass
CAS# 33309-88-5. 5G. Bis(tricyclohexylphosphine)palladium(0), 98%. MFCD01073796. Molecular Formula: C36H66P2Pd Molecular Weight: 667.28. Color/Form: off-white xtl. Stability: air sensitive, store cold. Strem#: 46-0260 www.strem.com/catalog/v3/46-0260/
Catalog No. 50-195-9577 Supplier Strem, An Ascensus Company Supplier No. 4602605G
CAS# 33309-88-5. 5G. Bis(tricyclohexylphosphine)palladium(0), 98%. MFCD01073796. Molecular Formula: C36H66P2Pd Molecular Weight: 667.28. Color/Form: off-white xtl. Stability: air sensitive, store cold. Strem#: 46-0260 www.strem.com/catalog/v3/46-0260/