Learn More
CPC Scientific H-Trp-Trp-Gly-Lys-Lys-Tyr-Arg-Ala-Ser-Lys-Leu-Gly-Leu-Ala-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific COMA001A
Encompass
(Trp63,Trp64)-C3a (63-77), synthetic superagonist analog of complement 3a, was 12-15 times more active than natural C3a.ONE-LETTER SEQUENCE: WWGKKYRASKLGLARMOLECULAR FORMULA: C86H134N26O18MOLECULAR WEIGHT:1820.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [130154-64-2], RESEARCH AREA: InflammationREFERENCES: J.A.Ember et al., Biochemistry, 30, 3603 (1991)
Catalog No. 50-210-9129