Learn More
CPC Scientific H-Ala-Ser-His-Leu-Gly-Leu-Ala-Arg-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific COMA002B
Encompass
The octapeptide C3a (70-77) is a C-terminal fragment of the C3a, has anaphylatoxin activity. This peptide interacts firectly with human lymphocyte.ONE-LETTER SEQUENCE: ASHLGLARMOLECULAR FORMULA: C35H61N13O10MOLECULAR WEIGHT:823.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [63555-63-5], RESEARCH AREA: InflammationREFERENCES: D.G.Payan et al., J. Exp. Med., 156, 756 (1982); E.L.Morgan et al., J. Immunol., 131, 2258 (1983); C.G.Unson et al., Biochemistry, 23, 585 (1984)
Catalog No. 50-210-9132