Learn More
Hycult Biotech Inc Skip to the beginning of the images galleryC3a/C3a des Arg, Human, mAb 2991, 500 ug
SDP
Supplier:  Hycult Biotech Inc HM2074500UG
Encompass
The monoclonal antibody 2991 recognizes the neo-epitope of human C3a/C3a-desArg that is formed upon cleavage of the complement protein C3 into C3a and C3b. The complement system is an important part of the humoral response in innate immunity, consisting of three different pathways. The third complement component, C3, is central to the classical, alternative and lectin pathways of complement activation.
Catalog No. NC1372578