Learn More
Hycult Biotech Inc C3A Human ELISA Kit, 2 x 96 detections, Sandwich ELISA, 100 μl/well, Minimum detectable concentration 31.3 pg/ml
SDP
Supplier:  Hycult Biotech Inc HK35402
Encompass
C3A Human ELISA Kit, 2 x 96 detections, Sandwich ELISA, 100 μl/well, Minimum detectable concentration 31.3 pg/ml
Catalog No. NC1489626