Learn More
Complement Technology Inc C3a Anaphylatoxin, 50 μg/vial, frozen liquid, 0.5 mg/mL, unglycosylated, >97% purity
Supplier: Complement Technology Inc A118
C3a Anaphylatoxin is a natural human C3a anaphylatoxin, which plays a vital role in mediating inflammatory responses in the body. This product is provided as a frozen liquid, with a concentration of 0.5 mg/mL and comes in quantities of 50 μg per vial. It is specifically prepared from normal human serum through the cleavage of the human C3 protein, ensuring a high level of purity and effectiveness in research applications.
- Source from normal human serum
- Preparation involves cleavage of human C3 protein
- Unglycosylated polypeptide consisting of 77 amino acids
- Molecular weight of 9,089 daltons (calculated)
- Concentration of 0.5 mg/mL
- Formulated as a frozen liquid
- Purity exceeds 97% by SDS-PAGE
- Stored at -70 °C or below; avoid freeze/thaw cycles
- Contains no preservatives
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.