Learn More
Enzo Life Sciences C3a-receptor Recombinant monoclonal antibody (3G7) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4040200
Encompass_Preferred
Clone: 3G7. Isotype: IgGκ. Immunogen: The 2e loop of the C3a receptor aa 185-193. Species reactivity: Human. Applications: ELISA, Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2931