Learn More
Abcam Human Complement C3b ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1954611X96TESTS
Encompass_Preferred
Human Complement C3b ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C3b in Human in Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1222929