Learn More
Diagnostic Biosystems C3C-FITC C3c Complement-FITC
Supplier:  Diagnostic Biosystems F00301
Encompass_Preferred
This antibody reacts with the human C3c complement and the C3c part of C3 and C3b. It may cross react with the C3c complement from cat, cow, dog, goat, guinea pig, kangaroo, mink, mouse, rat, sheep, and swine. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may be also used for other applications.
Catalog No. 50-240-7615