Learn More
CPC Scientific H-Lys-Asn-Arg-Trp-Glu-Asp-Pro-Gly-Lys-Gln-Leu-Tyr-Asn-Val-Glu-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific COMD001A
Encompass
P16, a 16-amino acid peptide sequence of human C3d, regulated in vitro and in vivo tyrosine phosphorylations of the pp100 and pp105 and in vitro proliferation of human B lymphocytes, depending on the stage of cell differentiation.ONE-LETTER SEQUENCE: KNRWEDPGKQLYNVEAMOLECULAR FORMULA: C86H131N25O27MOLECULAR WEIGHT:1947.14STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [99027-06-2], RESEARCH AREA: InflammationREFERENCES: R.Frade et al., Biochem. Biophys. Res. Commun., 188, 833 (1992)
Catalog No. 50-210-9133