Learn More
Electron Microscopy Sciences C3E Wipes, 9x9"
SDP
Catalog No. 50949373
Encompass_Preferred
These wipes are made from 100% synthetic fiber and spunlaced to provide excellent sorptive capacity. These durable wipes are exceptionally clean and have excellent chemical resistance, making them ideal for use with solvents. Recommended for use in ISO Class 5 Cleanroom applications.
This item is not returnable. View return policy
Catalog No. 50-949-373 Supplier Electron Microscopy Sciences Supplier No. 71762
These wipes are made from 100% synthetic fiber and spunlaced to provide excellent sorptive capacity. These durable wipes are exceptionally clean and have excellent chemical resistance, making them ideal for use with solvents. Recommended for use in ISO Class 5 Cleanroom applications.