Learn More
Aviva Systems Biology Corp C3orf10 Antibody - middle region (ARP51212_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51212_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human C3orf10. Alternate names: HSPC300, MDS027, hHBrk1, C3orf10ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-53654