Learn More
Aviva Systems Biology Corp NEPRO Antibody - middle region (ARP44752_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44752_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C3orf17. Alternate names: DKFZP434F2021, NET17ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58891