Learn More
Aviva Systems Biology Corp C3orf22 Antibody - middle region (ARP53027_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53027_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf22. It was validated on Western Blot. . Alternate names: MGC34728ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-285-795