Learn More
Aviva Systems Biology Corp C3orf49 Antibody - middle region (ARP53537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf49. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C3orf49. Alternate names: MGC17310ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-285-848