Learn More
Aviva Systems Biology Corp C3orf59 Antibody - C-terminal region (ARP53369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf59. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human C3orf59. Alternate names: C3orf59ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-830