Learn More
Aviva Systems Biology Corp C3orf62 Antibody - middle region (ARP55903_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55903_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf62. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C3orf62. Alternate names: FLJ43654, MGC23381, MGC61663, MGC62079ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61657