Learn More
Aviva Systems Biology Corp C3orf67 Antibody - N-terminal region (ARP50865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf67. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human C3orf67. Alternate names: FLJ42117, FLJ42930, MGC48416ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-546