Learn More
Cayman Chemical C4 Ceramide (d18:1/4:0), Purity: ≥95%, CAS: 74713-58-9, 50 mg
SDP
Catalog No. 502964084
Encompass_Preferred
A bioactive sphingolipid; inhibits IL-4 production by 16% in PMA-stimulated EL4 T cells at 10 μM; cytotoxic to SK-BR-3 and MCF-7/Adr breast cancer cells (IC50s = 15.9 and 19.9 μM, respectively); increases maturation and stability of F508del-CFTR proteins, enhances cAMP-activated chloride secretion, and suppresses secretion of IL-8 in primary epithelial cells isolated from patients with cystic fibrosis
Catalog No. 50-296-4084 Supplier Cayman Chemical Supplier No. 2252850 MG
A bioactive sphingolipid; inhibits IL-4 production by 16% in PMA-stimulated EL4 T cells at 10 μM; cytotoxic to SK-BR-3 and MCF-7/Adr breast cancer cells (IC50s = 15.9 and 19.9 μM, respectively); increases maturation and stability of F508del-CFTR proteins, enhances cAMP-activated chloride secretion, and suppresses secretion of IL-8 in primary epithelial cells isolated from patients with cystic fibrosis