Learn More
YMC America, Inc C4-HG col 120A 15um 250x30mm
SDP
Catalog No. 5000878323
Encompass
YMC-Pack C4 is ideal for analyzing mixtures with a wide range of component polarity This is due to the C4 bonded phase exhibiting shorter analysis time while still maintaining high resolution when compared to longer-chain bonded chemistries YMC-Pack C4 (Butyl) columns are available with three pore sizes to accommodate optimum separations of virtually any molecular-weight compound The 120 phase is used extensively with non-polar peptides which are too strongly retained on C8 and C18 The 200 and 300 phase columns effectively resolve many classes of proteins and biopolymers
Catalog No. 50-008-78323 Supplier YMC America, Inc Supplier No. BUG12S162530WT
YMC-Pack C4 is ideal for analyzing mixtures with a wide range of component polarity This is due to the C4 bonded phase exhibiting shorter analysis time while still maintaining high resolution when compared to longer-chain bonded chemistries YMC-Pack C4 (Butyl) columns are available with three pore sizes to accommodate optimum separations of virtually any molecular-weight compound The 120 phase is used extensively with non-polar peptides which are too strongly retained on C8 and C18 The 200 and 300 phase columns effectively resolve many classes of proteins and biopolymers