Learn More
ATCC C-4 I; Cervical Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL1594
Encompass
C-4 I is a cell line exhibiting epithelial morphology that was isolated from the cervix of a White, 41-year-old, female patient with carcinoma. This cell line was deposited N Auersperg and can be used in cancer, infectious disease and sexually transmitted disease research.
Catalog No. 50-238-1817