Learn More
Aviva Systems Biology Corp C4B Antibody - N-terminal region (ARP41841_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41841_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C4B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C4B. Alternate names: C4A, C4A13, C4A91, C4B1, C4B12, C4B2, C4B3, C4B5, C4F, CH, CO4, CPAMD3, MGC164979ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58015