Learn More
Aviva Systems Biology Corp C4BPA Antibody - middle region (ARP41420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C4BPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C4BPA. Alternate names: C4BP, PRPApplicationWB. Species reactivity: Human
Catalog No. 50-014-57840