Learn More
Diagnostic Biosystems C4C-FITC C4c Complement-FITC
Supplier:  Diagnostic Biosystems F005
Encompass_Preferred
This antibody reacts with human C4, C4b, and C4c but does not react with the C4d epitope. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7370