Learn More
Aviva Systems Biology Corp C4orf20 Antibody - middle region (ARP57217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C4orf20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C4orf20. Alternate names: FLJ11200, UFSP2, C4orf20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62108