Learn More
Aviva Systems Biology Corp CFAP299 Antibody - N-terminal region (ARP55538_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55538_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C4orf22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C4orf22. Alternate names: MGC35043ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-61533