Learn More
Aviva Systems Biology Corp C4orf33 Antibody - C-terminal region (ARP51908_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51908_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C4orf33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C4orf33. Alternate names: FLJ33703ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60643