Learn More
AdipoGen N-Pentanoyl-L-homoserine lactone (10 mg)
SDP
Supplier:  AdipoGen CDXP0486M010
Encompass_Preferred
Chemical. CAS: 148497-11-4. Formula: C9H15NO3. MW: 185.2. Synthetic. N-Pentanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. This regulatory process manifests itself with a variety of phenotypes including biofilm formation and virulence factor production. Regulation of bacterial quorum sensing signaling systems to inhibit pathogenesis represents a new approach to antimicrobial therapy in the treatment of infectious diseases.
Catalog No. 50-205-4614