Learn More
GENIE GROUP FLIR C5 Thermal Camera
Supplier:  GENIE GROUP 894010202
Encompass_Preferred
With the FLIR C5 in your pocket you’ll be ready anytime to find hot fuses, air leaks, plumbing issues, and more. Identifying hidden problems is easy with the 160 x 120 (19,200 pixels) true thermal imager, MSX (Multi-Spectral Dynamic Imaging), 5-megapixel visual camera, and LED flashlight. The C5 directly uploads and stores your images to the FLIR Ignite™ cloud, where you can organize and back up files. Then instantly share images with customers or create professional reports that document the problem and required repairs.
Catalog No. NC1893870