Learn More
Alkali Scientific Alkali Scientific™ C5000-6H-PT Sprint™ Portable Kit, 115V
SDP
Catalog No. 503029531
Encompass_Preferred
The Sprint™ 6H is a compact centrifuge that includes a six position, swing-out rotor. Prior to centrifugation, tubes are loaded vertically. Once the centrifuge starts, tubes swing to a horizontal position, providing the flat-line separation that is commonly preferred for further analysis. Sprint™ 6H Portable kit, 115V  
Catalog No. 50-302-9531 Supplier Alkali Scientific Supplier No. RS7532
The Sprint™ 6H is a compact centrifuge that includes a six position, swing-out rotor. Prior to centrifugation, tubes are loaded vertically. Once the centrifuge starts, tubes swing to a horizontal position, providing the flat-line separation that is commonly preferred for further analysis. Sprint™ 6H Portable kit, 115V  